ContactUsLinkComponent
Mortars, porcelain, with spout, glazed outside, rough inside, without pestle, 83 mm Ø, 50 mm height, 110 ml, 2 pcs./carton, 211a/2 1 * 2 items
Catalog # MARI6835112
Supplier:  MARIENFELD
Mortars, porcelain, with spout, glazed outside, rough inside, without pestle, 83 mm Ø, 50 mm height, 110 ml, 2 pcs./carton, 211a/2 1 * 2 items
Catalog # MARI6835112
Supplier:  MARIENFELD
Frequently Bought Together