Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Hydroxy-4-methyl-2-pentanone 95%

4-Hydroxy-4-methyl-2-pentanone 95%

Bottle, 500 g
Catalog # APOSOR924093-500G
Supplier:   Apollo Scientific
4-Hydroxy-4-methyl-2-pentanone 95%
Catalog # APOSOR924093-500G
Supplier:  Apollo Scientific
CAS Number:  123-42-2
4-Hydroxy-4-methyl-2-pentanone 95%

Bottle, 500 g

CAS Number:  123-42-2
Price
268,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Diacetone alcohol
  • CAS Number:123-42-2
  • Formula:(CH₃)₂C(OH)CH₂COCH₃
  • MDL Number:MFCD00004471
  • Molecular Weight:116.16 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.