Curcumin 97%
Bottle, 25 g
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
- CAS Number:458-37-7
- Formula:C₂₁H₂₀O₆
- MDL Number:MFCD00008365
- Molecular Weight:368.39 g/mol
- Melting Pt:170…175 °C
- Storage Temperature:Freezer
- EINECS:207-280-5
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:25 g



