ContactUsLinkComponent
Human CCR5, Flag,His Tag CC5-H52D1 is expressed from human 293 cells HEK293. It contains AA Asp 2 - Arg 223 Glu 227 - Arg 319 Acces sion P51681-1 C58Y, G163N, A233D, K303E. 1 * 500 µG
Catalog # ACRBCC5-H52D1500UG
Supplier:  ACROBIOSYSTEMS
Human CCR5, Flag,His Tag CC5-H52D1 is expressed from human 293 cells HEK293. It contains AA Asp 2 - Arg 223 Glu 227 - Arg 319 Acces sion P51681-1 C58Y, G163N, A233D, K303E. 1 * 500 µG
Catalog # ACRBCC5-H52D1500UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together