Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat (N-plan) oil phase Ph3; 1,25 NA, 0,15 mm
Catalog # ECHLJMAG1009
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG1009
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat (N-plan) oil phase Ph3; 1,25 NA, 0,15 mm
Price
1 670,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    100×
  • Description:
    Plan achromat (N-plan) oil phase Ph3; 1,25 NA
  • Seq:
    28
  • Working distance:
    0,15 mm