Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium hydroxide, pellets NF, Multi-Compendial

Sodium hydroxide, pellets NF, Multi-Compendial

PE pail, screw top, 12 kg
Catalog # 3717-07
Corrosive
Supplier:   Avantor
Sodium hydroxide, pellets NF, Multi-Compendial
Catalog # 3717-07
Supplier:  Avantor
CAS Number:  1310-73-2
Sodium hydroxide, pellets NF, Multi-Compendial
Sodium hydroxide, pellets NF, Multi-Compendial

PE pail, screw top, 12 kg

CAS Number:  1310-73-2
Price
2 066,35 €
Log in for availability
1 * 12 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:1390 °C
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:1823
  • ADR:8,II

Documents

Specifications

Specification Test Results

  • NF - Identification A (Sodium):
    Passes Test
  • NF - Identification B (pH):
    ≥11.0
  • NF - Insoluble Substances and Organic Matter:
    Passes Test
  • NF - Potassium (K):
    ≤0.5 %
  • NF - Carbonate (as Na₂CO₃):
    ≤3.0 %
  • NF - Total Alkali:
    95.0 - 100.5 %
  • NF - Content of Sodium:
    54.0 - 59.8 %
  • ChP -Assay:
    97.0 - 100.5 %
  • ChP -Sodium carbonate:
    2.0% max
  • ChP -Identification:
    Passes Test
  • ChP -Alkalinity:
    Passes Test
  • ChP -Clarity and color of solution :
    Passes Test
  • ChP -Chloride :
    0.005 % max
  • ChP -Sulfate :
    0.015 % max
  • ChP -Potassium :
    Passes Test
  • ChP -Aluminum and iron :
     0.1% max
  • ChP -Iron :
    0.001% max
  • ChP -Heavy metal :
    20 ppm max
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Carbonate (as Na₂CO₃):
    ≤2.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤200 ppm
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Assay (NaOH):
    97.0 - 100.5 %
  • JP - Assay:
    95.0 - 101.0 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤0.050 %
  • JP - Heavy Metals (as Pb):
    ≤30 ppm
  • JP - Potassium (K):
    Passes Test
  • JP - Carbonate (as Na₂CO₃):
    ≤2.0 %
  • JP - Mercury (Hg):
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤5

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.