Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium hydroxide, pellets NF, Multi-Compendial

Sodium hydroxide, pellets NF, Multi-Compendial

Poly bottle, 500 g
Catalog # 3717-01
Corrosif
Supplier:   Avantor
Sodium hydroxide, pellets NF, Multi-Compendial
Catalog # 3717-01
Supplier:  Avantor
CAS Number:  1310-73-2
Sodium hydroxide, pellets NF, Multi-Compendial
Sodium hydroxide, pellets NF, Multi-Compendial

Poly bottle, 500 g

CAS Number:  1310-73-2
Price
158,70 €
Connexion Pour la disponibilité
1 * 500 g
Product Details & Documents
Synonymes:  Caustic soda
  • Numéro CAS:1310-73-2
  • Formule:NaOH
  • Numéro MDL:MFCD00003548
  • Poids moléculaire:40 g/mol
  • Point d'ébullition:1390 °C
  • Point de fusion:323 °C
  • Densité:2.13 g/cm³ (20 °C)
  • Température de stockage:Ambient
  • UN:1823
  • ADR:8,II

Documents

Spécifications

Résultats des tests de spécification

  • NF - Identification A (Sodium):
    Passes Test
  • NF - Identification B (pH):
    ≥11.0
  • NF - Insoluble Substances and Organic Matter:
    Passes Test
  • NF - Potassium (K):
    ≤0.5 %
  • NF - Carbonate (as Na₂CO₃):
    ≤3.0 %
  • NF - Total Alkali:
    95.0 - 100.5 %
  • NF - Content of Sodium:
    54.0 - 59.8 %
  • ChP -Assay:
    97.0 - 100.5 %
  • ChP -Sodium carbonate:
    2.0% max
  • ChP -Identification:
    Passes Test
  • ChP -Alkalinity:
    Passes Test
  • ChP -Clarity and color of solution :
    Passes Test
  • ChP -Chloride :
    0.005 % max
  • ChP -Sulfate :
    0.015 % max
  • ChP -Potassium :
    Passes Test
  • ChP -Aluminum and iron :
     0.1% max
  • ChP -Iron :
    0.001% max
  • ChP -Heavy metal :
    20 ppm max
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Carbonate (as Na₂CO₃):
    ≤2.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤200 ppm
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Assay (NaOH):
    97.0 - 100.5 %
  • JP - Assay:
    95.0 - 101.0 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤0.050 %
  • JP - Heavy Metals (as Pb):
    ≤30 ppm
  • JP - Potassium (K):
    Passes Test
  • JP - Carbonate (as Na₂CO₃):
    ≤2.0 %
  • JP - Mercury (Hg):
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤5

Spécifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.