ContactUsLinkComponent
membrane pump and 3,3kW steam generator for the operation of device without connection to demineralised water, tap water and drainage 1 * 1 items
Catalog # SHPS209999999100
Supplier:  SHP Steriltechnik
membrane pump and 3,3kW steam generator for the operation of device without connection to demineralised water, tap water and drainage 1 * 1 items
Catalog # SHPS209999999100
Supplier:  SHP Steriltechnik
Frequently Bought Together