ContactUsLinkComponent
The Vacuum Blotter is built to quickly and efficiently transfer DNA or RNA onto suitable membrane such as Positive Nylon membrane. Vacuum bloting gives excellent results provided vacuum pressure is precise and stable during the blotting procedure. Ga 1 * 5 items
Catalog # 11INSB8302
Supplier:  MP Biomedicals
The Vacuum Blotter is built to quickly and efficiently transfer DNA or RNA onto suitable membrane such as Positive Nylon membrane. Vacuum bloting gives excellent results provided vacuum pressure is precise and stable during the blotting procedure. Ga 1 * 5 items
Catalog # 11INSB8302
Supplier:  MP Biomedicals
Frequently Bought Together