Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Plan fluorite water dipping; 0,5 NA, 3,5 mm
Catalog # ECHLJMAG0218
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # ECHLJMAG0218
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Plan fluorite water dipping; 0,5 NA, 3,5 mm
Price
2 220,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    20×
  • Description:
    Plan fluorite water dipping; 0,5 NA
  • Seq:
    22
  • Working distance:
    3,5 mm