Orders LinkContactUsLinkComponent
VWR®, Cleanroom clogs
VWR®, Cleanroom clogs
XL (EU: 43/44), White
Catalog # 114-8984
Supplier:   VWR Collection
VWR®, Cleanroom clogs
Catalog # 114-8984
Supplier:  VWR Collection
VWR®, Cleanroom clogs
VWR®, Cleanroom clogs
XL (EU: 43/44), White
Price
168,00 €
Log in for availability
1 * 1 PAIRE
Product Details & Documents
VWR® cleanroom clogs provide comfort and durability.
  • Anti-static
  • Autoclavable
  • Smooth surface for day long comfort
  • Special conical design prevents spilled liquids from entering clog
  • Heel strap
  • Non slip sole
  • Anatomic heel basin
They have been designed with foot comfort in mind. Smooth surface and aeration holes on both sides provide day extended comfort. Special conical design prevents spilled liquids from entering inside the clog.
These shoes are suitable for autoclave sterilisation. They can also be cleaned in 100 °C with domestic detergents, liquid disinfecting solutions, EO, or formaldehyde.
Ideal for use with VWR® cleanroom disposable or launderable socks.
Ordering information: Clogs can be ordered by the pair or per case.
Packaging: Packed six pairs per case.
Specifications
  • Shoe size:
    XL (EU: 43/44)
  • Colour:
    White