Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Fast extract DNA solution

VWR®, Fast extract DNA solution

VWR® fast extract DNA solution, 8 min, 500 reactions, 5×10 ml, 500 Tests
Catalog # 733-2877
Supplier:   VWR Chemicals
VWR®, Fast extract DNA solution
Catalog # 733-2877
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Fast extract DNA solution
VWR®, Fast extract DNA solution

VWR® fast extract DNA solution, 8 min, 500 reactions, 5×10 ml, 500 Tests

CAS Number:  
Price
269,00 €
Log in for availability
1 * 500 Tests
Product Details & Documents

The fast extract DNA solution provides rapid and efficient extraction of PCR-ready DNA from mammalian and plant tissues. DNA is ready in eight minutes. The one-reagent protocol is divided into two simple heating steps, which can be directly followed by PCR analysis, such as screening and genotyping.

  • One-reagent set-up
  • Rapid eight-minute protocol
  • PCR-ready DNA
  • DNA extracts from mammalian tissues, plant materials, and bacteria
  • Non-toxic reagents
  • Automation-friendly

DNA can be extracted from many different sample types e.g. mouse tails or ears, saliva, bacteria, plant material, and mammalian tissues . The one-reagent DNA extraction set-up is easily scaled and can be conducted by robotic automation platforms. Depending on the sample size, the DNA extraction can be performed in PCR tubes or in 1,5 ml tubes, using either a thermocycler or a heating block.

Prep size: 100 µl
Method/format: DNA extraction

Specifications

Specifications

  • Pk:
    500 Tests
  • Description:
    VWR® fast extract DNA solution, 8 min, 500 reactions
  • Volume:
    5×10 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.