Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

VWR®, Fast extract DNA solution

VWR® fast extract DNA solution, 8 min, 100 reactions, 1×10 ml, 100 Tests
Catalog # 733-2876
Supplier:   VWR Chemicals
VWR®, Fast extract DNA solution
Catalog # 733-2876
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Fast extract DNA solution
VWR®, Fast extract DNA solution

VWR® fast extract DNA solution, 8 min, 100 reactions, 1×10 ml, 100 Tests

CAS Number:  
Price
59,30 €
Log in for availability
1 * 1 EA
Product Details & Documents

The fast extract DNA solution provides rapid and efficient extraction of PCR-ready DNA from mammalian and plant tissues. DNA is ready in eight minutes. The one-reagent protocol is divided into two simple heating steps, which can be directly followed by PCR analysis, such as screening and genotyping.

  • One-reagent set-up
  • Rapid eight-minute protocol
  • PCR-ready DNA
  • DNA extracts from mammalian tissues, plant materials, and bacteria
  • Non-toxic reagents
  • Automation-friendly

DNA can be extracted from many different sample types e.g. mouse tails or ears, saliva, bacteria, plant material, and mammalian tissues . The one-reagent DNA extraction set-up is easily scaled and can be conducted by robotic automation platforms. Depending on the sample size, the DNA extraction can be performed in PCR tubes or in 1,5 ml tubes, using either a thermocycler or a heating block.

Prep size: 100 µl
Method/format: DNA extraction

Specifications

Specifications

  • Pk:
    100 Tests
  • Description:
    VWR® fast extract DNA solution, 8 min, 100 reactions
  • Volume:
    1×10 ml

Specifications