ContactUsLinkComponent
THESE STAINLESS STEEL FINE-POINT FORCEPS ARE 4.25 IN (110 MM) IN LENGTH, ARE STRAIGHT, NON-SERRATED AND ARE MADE FROM 316L LOW CARBO N AUSTENITIC STEEL. THE TWEEZERS ARE NON-MAGNETIZABLE AND HAVE GOOD CORROSION RESISTANCE TO MOST CHEMICALS, SALTS AND 1 * 1 items
Catalog # PERCL9001976
Supplier:  PerkinElmer
THESE STAINLESS STEEL FINE-POINT FORCEPS ARE 4.25 IN (110 MM) IN LENGTH, ARE STRAIGHT, NON-SERRATED AND ARE MADE FROM 316L LOW CARBO N AUSTENITIC STEEL. THE TWEEZERS ARE NON-MAGNETIZABLE AND HAVE GOOD CORROSION RESISTANCE TO MOST CHEMICALS, SALTS AND 1 * 1 items
Catalog # PERCL9001976
Supplier:  PerkinElmer
Frequently Bought Together