ContactUsLinkComponent
THE AUTOSAMPLER HAS A 3-CHANNEL PERISTALTIC PUMP THAT ACCEPTS BELOW LISTED TUBES WHEN RUNNING INORGANIC ACIDS. 2 - 2 MM ID PHARMX, B LACK TABS, RINSE 1 - 3.2MM ID PHARMX, WHITE TABS, WASTE 1 * 1 items
Catalog # PERCN0831030
Supplier:  PerkinElmer
THE AUTOSAMPLER HAS A 3-CHANNEL PERISTALTIC PUMP THAT ACCEPTS BELOW LISTED TUBES WHEN RUNNING INORGANIC ACIDS. 2 - 2 MM ID PHARMX, B LACK TABS, RINSE 1 - 3.2MM ID PHARMX, WHITE TABS, WASTE 1 * 1 items
Catalog # PERCN0831030
Supplier:  PerkinElmer
Frequently Bought Together