CONCENTRATION OF ELEMENT INCLUDES: ELEMENT CONCENTRATION CADMIUM 200 µG/ML CHROMIUM 200 µG/ML IRON 200 µG/ML LITHIUM 200 µG/ML MAGNE SIUM 200 µG/ML LEAD 200 µG/ML COPPER 1 µG/ML NITRIC ACID 1% 1 * 1 items
Catalog # PERCN8162302
Supplier: PerkinElmer
Frequently Bought Together