Orders LinkContactUsLinkComponent
CloneMatrix
CloneMatrix
CloneMatrix CHO (1×40 ml), 1 Bottle
Catalog # MLDVK8530
Supplier:   Molecular Devices
CloneMatrix
Catalog # MLDVK8530
Supplier:  Molecular Devices
CAS Number:  
CloneMatrix
CloneMatrix CHO (1×40 ml), 1 Bottle
CAS Number:  
Price
637,00 €
Log in for availability
1 * 1 Fl
Product Details & Documents
CloneMatrix is a 2,5X concentrate that serves as a matrix to produce semi-solid media for hybridoma, myeloma, CHO, HEK, and mESC.
Specifications
Specifications
  • Pk:
    1 Bottle
  • Description:
    CloneMatrix CHO (1×40 ml)
Specifications