Reagecon BY5 Colour Reference Solution according to European Pharmacopoeia (EP) 1 * 100 mL
Catalog # REGEEPBY105
Supplier: REAGECON
Reagecon BY5 Colour Reference Solution according to European Pharmacopoeia (EP) 1 * 100 mL
Catalog # REGEEPBY105
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.