Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sampling scoops, LaboPlast®

Standard, 2000 ml, 381 mm, White
Catalog # 331-0365
Supplier:   BURKLE
Sampling scoops, LaboPlast®
Catalog # 331-0365
Supplier:  BURKLE
Sampling scoops, LaboPlast®
Sampling scoops, LaboPlast®

Standard, 2000 ml, 381 mm, White

Price
97,10 €
Log in for availability
1 * 10 ST
Product Details & Documents

PS, white. These scoops are ideal for the sampling of powder, granules and pastes. They are also available with a transparent lid to close it immediately after the sampling procedure to secure a clean and contamination-free sample.

  • Individually packed
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines

Due to the special handle construction and the slightly angled edge the scoop stands horizontal and can be laid down without the material trickling out.

Specifications
  • Type:
    Standard
  • Capacity:
    2000 ml
  • Colour:
    White
  • Length:
    381 mm
  • Seq:
    14
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.