Order Entry
Belgium
Orders LinkContactUsLinkComponent
cDNA synthesis kits, AffinityScript

cDNA synthesis kits, AffinityScript

AffinityScript One-Step RT-PCR Kit, Reduced PCR cycling times and a one-tube, master mix format. Combined performance of Herculase II fusion DNA polymerase and the ArchaeMaxx PCR enhancing factor, delivering superior PCR yield and reliability. Robust performance for difficult and GC-rich targets. Optimised for target lengths up to 9,5 KB, 100 reactions, 1
Catalog # AGLS600188
Supplier:   AGILENT
cDNA synthesis kits, AffinityScript
Catalog # AGLS600188
Supplier:  AGILENT
CAS Number:  
cDNA synthesis kits, AffinityScript

AffinityScript One-Step RT-PCR Kit, Reduced PCR cycling times and a one-tube, master mix format. Combined performance of Herculase II fusion DNA polymerase and the ArchaeMaxx PCR enhancing factor, delivering superior PCR yield and reliability. Robust performance for difficult and GC-rich targets. Optimised for target lengths up to 9,5 KB, 100 reactions, 1

CAS Number:  
Price
422,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Provides the highest efficiency of conversion of RNA to cDNA, fully optimised for qPCR applications.

  • Optimised for real-time quantitative PCR (qPCR) application;Produces cDNA in 15 min with linear detection from 3 pg to 3 µg total RNA
  • Creates up to 12 kb cDNA in 25 min or less
  • Available as a first-strand synthesis kit and a two-step QRT-PCR master mix format

The AffinityScript qPCR cDNA Synthesis Kit offers efficient conversion of RNA to cDNA. Fully optimised for real-time quantitative PCR (qPCR) applications, the kit utilises our qPCR-grade AffinityScript Multiple Temperature Reverse Transcriptase. It includes a buffer system optimised for real-time qPCR.

The streamlined protocol produces cDNA in 15 minutes with linear detection from 3 pg to 3 µg of total RNA. The easy-to-use process generates up to 12 kb cDNA in 25 min or less. Available as a first-strand synthesis kit and a two-step QRT-PCR master mix format, using SYBR Green or probe-based detection.

The AffinityScript Multiple Temperature cDNA synthesis kit contains Agilent’s popular, highly thermostable, qPCR-grade AffinityScript Multiple Temperature Reverse Transcriptase for robust yields, from 37 to 60 °C, with nearly full activity between 45 to 55 °C. With the AffinityScript multiple temperature cDNA synthesis kit, Agilent offers enzymes with a broad thermal profile.

The Agilent AffinityScript Multiple Temperature Reverse Transcriptase (RT) system is optimised to produce high cDNA yields over the broadest temperature range. During analysis the multi-temp RT device also ensures high affinity to primer-template complexes. cDNA can be synthesised in amounts up to 20 kb.

The system’s multi-temperature capability allows you to change RT reaction temperature without having to change the reverse transcriptase, for a faster and more efficient analysis. In addition, when amplifying low RNA input amounts and resolving RNA secondary structures it delivers high cDNA yields, and can operate effectively at temperatures up to 55 °C.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Pk:
    1
  • Description:
    AffinityScript One-Step RT-PCR Kit
  • Details:
    Reduced PCR cycling times and a one-tube, master mix format. Combined performance of Herculase II fusion DNA polymerase and the ArchaeMaxx PCR enhancing factor, delivering superior PCR yield and reliability. Robust performance for difficult and GC-rich targets. Optimised for target lengths up to 9,5 KB
  • No. of reactions:
    100 reactions

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.