Order Entry
Belgium
Orders LinkContactUsLinkComponent
DNase I

DNase I

5000 units, 1
Catalog # AGLS600032
Supplier:   AGILENT
DNase I
Catalog # AGLS600032
Supplier:  AGILENT
CAS Number:  
DNase I

5000 units, 1

CAS Number:  
Price
411,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Purified DNase I tested to be free of contaminating activity for the degradation of nucleic acids.

  • RNase free
  • Contains a mixture of four isoenzymes from bovine pancreas (10 units/µl)

DNase I, RNase Free catalyses the degradation of double-stranded DNA into oligonucleotides 1,2 and mononucleotides. This enzyme has been isolated as a mixture of four isoenzymes from bovine pancreas that cut preferentially next to pyrimidine nucleotides.

Reaction Conditions: 40 mM Tris-HCl (pH 7,5), 6 mM MgCl₂, 2 mM CaCl₂, 1 µg DNA and 1 unit enzyme in a 25 µl volume. Incubate 30 minutes at 37 °C.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Pk:
    1
  • Pk Info:
    5000 units
  • Cat. No.:
    AGLS600032
  • Form:
    Premixed
  • Formulation:
    10 mM Tris-HCl (pH 7,5), 10 mM CaCl₂, 10 mM MgCl₂ and 50 % glycerol (v/v). (Dilutions should be made in this buffer.)
  • Enzyme name:
    DNase I

Specifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.