Order Entry
Belgium
Orders LinkContactUsLinkComponent
DNase I

DNase I

1000 units, 1
Catalog # AGLS600031
Supplier:   AGILENT
DNase I
Catalog # AGLS600031
Supplier:  AGILENT
CAS Number:  
DNase I

1000 units, 1

CAS Number:  
Price
205,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Purified DNase I tested to be free of contaminating activity for the degradation of nucleic acids.

  • RNase free
  • Contains a mixture of four isoenzymes from bovine pancreas (10 units/µl)

DNase I, RNase Free catalyses the degradation of double-stranded DNA into oligonucleotides 1,2 and mononucleotides. This enzyme has been isolated as a mixture of four isoenzymes from bovine pancreas that cut preferentially next to pyrimidine nucleotides.

Reaction Conditions: 40 mM Tris-HCl (pH 7,5), 6 mM MgCl₂, 2 mM CaCl₂, 1 µg DNA and 1 unit enzyme in a 25 µl volume. Incubate 30 minutes at 37 °C.

 : For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Pk:
    1
  • Pk Info:
    1000 units
  • Cat. No.:
    AGLS600031
  • Form:
    Premixed
  • Formulation:
    10 mM Tris-HCl (pH 7,5), 10 mM CaCl₂, 10 mM MgCl₂ and 50 % glycerol (v/v). (Dilutions should be made in this buffer.)
  • Enzyme name:
    DNase I

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.