ContactUsLinkComponent
Latest version of SoftMax Pro 7 GxP for Dedicated Server Setup installation. Compatible with Windows 10 Professional/Enterprise. Includes 3-fold CD package 1) GxP Admin CD 2) SMP7 GxP CD 3) Software Validation CD and GxP User License certificate and 1 * 1 items
Catalog # MLDVSMP7XGXPSERVER
Supplier:  Molecular Devices
Latest version of SoftMax Pro 7 GxP for Dedicated Server Setup installation. Compatible with Windows 10 Professional/Enterprise. Includes 3-fold CD package 1) GxP Admin CD 2) SMP7 GxP CD 3) Software Validation CD and GxP User License certificate and 1 * 1 items
Catalog # MLDVSMP7XGXPSERVER
Supplier:  Molecular Devices
Frequently Bought Together