Waterproof alarm thermometer with immersion probe G1710-WPT2A Probe (permanently connected): With immersion probe, Ø3 mm, handle and cable shorttime 250°C, permanent 230°C Measuring input: Pt1000 (2-wire) Cable: Cable length 1 m silicone cable Displa 1 * 1 items
Catalog # GRSG481377
Supplier: Greisinger
Frequently Bought Together