ContactUsLinkComponent
CHROMABOND columns HLB (30 µm, fine grained) particle size: 30 µm volume: 6 mL, content of sorbent: 200 mg material: PP, with PE fi lter elements pack of 30 1 * 30 items
Catalog # MANA730926P30
Supplier:  MACHEREY-NAGEL
CHROMABOND columns HLB (30 µm, fine grained) particle size: 30 µm volume: 6 mL, content of sorbent: 200 mg material: PP, with PE fi lter elements pack of 30 1 * 30 items
Catalog # MANA730926P30
Supplier:  MACHEREY-NAGEL
Frequently Bought Together