Programmable Digital Muffle Furnace, Type FHP-14, with 4-side heating & program digital PID control system, temperature range: 300° - 1200°C, capacity: 14 Liter, internal: 250 x 350 x 160 mm, external: 530 x 580 x 620 mm, power: 4,0 kW,power supply: 1 * 1 items
Catalog # WITGDH.WFH23.P14
Supplier: witeg Labortechnik
Frequently Bought Together