ContactUsLinkComponent
Thermometer similar to ASTM 3 C, solid stem, white backed, -5+400:1C, gallium filled, immersion 76mm, 415x6-7,5mm, durable pigment, calibrated, with DKD-Certificate (DAkkS) with 5 test points 1 * 1 items
Catalog # AMARA300050GA/DKD5
Supplier:  Amarell
Thermometer similar to ASTM 3 C, solid stem, white backed, -5+400:1C, gallium filled, immersion 76mm, 415x6-7,5mm, durable pigment, calibrated, with DKD-Certificate (DAkkS) with 5 test points 1 * 1 items
Catalog # AMARA300050GA/DKD5
Supplier:  Amarell
Frequently Bought Together