ContactUsLinkComponent
Tube, Specimen Collection and Transport Tube Labcon SuperClear® 10 mL Cap with integral molded elastomeric seal Includes molded graduations and writing surface ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) IATA 95kPa certified to conform 1 * 500 items
Catalog # 3814-870-008-9
Supplier:  Labcon
undefined
Tube, Specimen Collection and Transport Tube Labcon SuperClear® 10 mL Cap with integral molded elastomeric seal Includes molded graduations and writing surface ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) IATA 95kPa certified to conform 1 * 500 items
Catalog # 3814-870-008-9
Supplier:  Labcon