Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Scroll vacuum pump

Silencer
Catalog # LBCP7587300
Supplier:   LABCONCO
Scroll vacuum pump
Catalog # LBCP7587300
Supplier:  LABCONCO
Scroll vacuum pump
Scroll vacuum pump

Silencer

Price
754,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Scroll Vacuum Pumps are low maintenence, corrosion resistent pumps that require no oil.

  • Scrolls be changed every 40000 hours of use
  • Eliminating the oil eliminates hydrocarbon contamination

Handle water vapor better than most types of pumps and make significantly less noise during operation. Works with FreeZone triad, 1, 2, 4, 5, 6, 8, 12 and 18 L freeze dryers.

Specifications
  • Description:
    Silencer
  • Seq:
    03
  • Cat. No.:
    LBCP7587300
  • Ultimate vacuum:
    7 x 10-3 mBar (Partial Pressure)
  • Dimensions:
    101,6L×25,4W×25,4H mm
  • Final vacuum:
    <7×10-3 mbar
  • Electrical: