SpectraTest ABS2 absorbance validation plate for SpectraMax M2, M2e, M3, M4, M5, M5e, i3, i3x, iD3, iD5, Plus 384, ABS, ABS Plus, and FlexStation 3. Includes validation plate, paper calibration certificate, software protocols and digital calibration 1 * 1 items
Catalog # MLDV0200-6191
Supplier: Molecular Devices
Frequently Bought Together