ContactUsLinkComponent
Tungsten carbide disposable blades TC65 are specially designed and manufactured to meet the stringent requireements of laboratories routinely sectioning hard and abrasive materials. The unique cutting edge treatment guarantees sections down to appr 1 * 5 items
Catalog # LEIH14021626379
Supplier:  LEICA HISTOLOGY
Tungsten carbide disposable blades TC65 are specially designed and manufactured to meet the stringent requireements of laboratories routinely sectioning hard and abrasive materials. The unique cutting edge treatment guarantees sections down to appr 1 * 5 items
Catalog # LEIH14021626379
Supplier:  LEICA HISTOLOGY
Frequently Bought Together