Hook2Loop Bungee tool tether, DBI-SALA®, 3M™
Hook2Loop Bungee tether, yellow, 6,8 kg capacity
N° de catalogue MMMA1500047
Fournisseur: 3M
Aller à :
Product Details & Documents
Bungee tool tether with self-locking carabiner.
- Adjusts to virtually any wrist size with hook and loop system
- Replaceable steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
- Elastic band helps ensure a comfortable fit
- Attached trigger can be used to quickly work with many hand tools
This is a very versatile extension that can be used with virtually any tool, such as spud wrenches, scaffold wrenches, adjustable wrenches, and more.
Designed for use in general industrial, oil and gas, mining, construction, and transportation industries.
Documents
3M™ Fall Protection Core Product Range 3M™ Fall Protection for Tools
Documentation produit
Spécifications
- Description:Hook2Loop Bungee tether, yellow, 6,8 kg capacity
- Seq:02
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.