Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat LWD; 0,4 NA, 2,8 mm
Catalog # ECHLJMAG0216
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG0216
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat LWD; 0,4 NA, 2,8 mm
Price
1 420,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    20×
  • Description:
    Plan achromat LWD; 0,4 NA
  • Seq:
    16
  • Working distance:
    2,8 mm