ContactUsLinkComponent
Recirculating Cooler Unichiller 070Tw with Pilot ONE -10...40 °C 1 * 1 items
Catalog # HUBE3016.0030.01
Supplier:  HUBER
Recirculating Cooler Unichiller 070Tw with Pilot ONE -10...40 °C 1 * 1 items
Catalog # HUBE3016.0030.01
Supplier:  HUBER
Frequently Bought Together