ContactUsLinkComponent
Recirculating Cooler Unichiller 100w with Pilot ONE -20...40 °C 1 * 1 items
Catalog # HUBE3059.0009.01
Supplier:  HUBER
Recirculating Cooler Unichiller 100w with Pilot ONE -20...40 °C 1 * 1 items
Catalog # HUBE3059.0009.01
Supplier:  HUBER
Frequently Bought Together