Selenium AAS Standards, (CRMs), Fluka™
Sélénium , Se, 1000 mg/L, 2% HNO₃, Plastic bottle, 500 mL
Catalog # HONCE4067-500ML
Supplier: Honeywell Chemicals
Selenium AAS Standards, (CRMs), Fluka™
Catalog # HONCE4067-500ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifications
Specifications
- Concentration:1000 mg/L
- Matrice:2% HNO₃
- Type de conditionnement:Plastic bottle
- Cdt:500 mL
- Élement:Sélénium
- Symbole:Se
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



