Selenium ICP Standards, (CRMs), Fluka™
Selenium, Se, 10000 mg/L, 5% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE1181-100ML
Supplier: Honeywell Chemicals
Selenium ICP Standards, (CRMs), Fluka™
Catalog # HONCE1181-100ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifications
Specifications
- Concentration:10000 mg/L
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Selenium
- Symbol:Se



