Order Entry
Belgium
Orders LinkContactUsLinkComponent
Selenium ICP Standards, (CRMs), Fluka™

Selenium ICP Standards, (CRMs), Fluka™

Selenium, Se, 1000 mg/L, 2% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE1054-100ML
Supplier:   Honeywell Chemicals
Selenium ICP Standards, (CRMs), Fluka™
Catalog # HONCE1054-100ML
CAS Number:  
Selenium ICP Standards, (CRMs), Fluka™

Selenium, Se, 1000 mg/L, 2% HNO₃, Plastic bottle, 100 mL

CAS Number:  
Price
79,20 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST

Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).

Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Selenium
  • Symbol:
    Se

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.