ContactUsLinkComponent
Dino-Lite model AF4915ZT,20-220x magnification, compatible with the WF-20 Wireless adapter (not included), USB 2.0, Enhanced 1.3MP EDGE SENSOR, Integrated Polarizer (anti-reflection), Extensive measurement functionality, Flexible LED control (FLC) , 1 * 1 items
Catalog # IDCPAF4915ZT
Dino-Lite model AF4915ZT,20-220x magnification, compatible with the WF-20 Wireless adapter (not included), USB 2.0, Enhanced 1.3MP EDGE SENSOR, Integrated Polarizer (anti-reflection), Extensive measurement functionality, Flexible LED control (FLC) , 1 * 1 items
Catalog # IDCPAF4915ZT
Frequently Bought Together