ContactUsLinkComponent
SpectraMax® Glow Luminescence (LUM) (384) Detection Cartridge for 384-well plates, utilizes a unique, well-isolating design that sig nificantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax® PARADIGM® and/or Spe t 1 * 1 items
Catalog # MLDV0200-7015
Supplier:  Molecular Devices
SpectraMax® Glow Luminescence (LUM) (384) Detection Cartridge for 384-well plates, utilizes a unique, well-isolating design that sig nificantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax® PARADIGM® and/or Spe t 1 * 1 items
Catalog # MLDV0200-7015
Supplier:  Molecular Devices
Frequently Bought Together