ContactUsLinkComponent
SpectraMax® Cisbio HTRF Detection Cartridge utilizes a high energy Xenon flash lamp for sensitive reading of HTRF reagents. Compati ble with SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. In addition, the standard dual emission design gives t t 1 * 1 items
Catalog # MLDV0200-7011
Supplier:  Molecular Devices
SpectraMax® Cisbio HTRF Detection Cartridge utilizes a high energy Xenon flash lamp for sensitive reading of HTRF reagents. Compati ble with SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. In addition, the standard dual emission design gives t t 1 * 1 items
Catalog # MLDV0200-7011
Supplier:  Molecular Devices
Frequently Bought Together