ContactUsLinkComponent
SpectraMax® PARADIGM® Multimode (MULTI) Detection Cartridge is optimized for 96- and 384-well plates and provides a broad array of s ingle-emission measurement capabilities for Fluorescence Intensity (FI),Time Resolved Fluorescence (TRF) and Lumines 1 * 1 items
Catalog # MLDV0200-7001
Supplier:  Molecular Devices
SpectraMax® PARADIGM® Multimode (MULTI) Detection Cartridge is optimized for 96- and 384-well plates and provides a broad array of s ingle-emission measurement capabilities for Fluorescence Intensity (FI),Time Resolved Fluorescence (TRF) and Lumines 1 * 1 items
Catalog # MLDV0200-7001
Supplier:  Molecular Devices
Frequently Bought Together