Orders LinkContactUsLinkComponent
RackScan
RackScan
RackScan s, code scanner for side reading of SBS racks and cryostorage boxes, accessory for the RackScan product family
Catalog # EPPE6002000.014
Supplier:   EPPENDORF
RackScan
Catalog # EPPE6002000.014
Supplier:  EPPENDORF
RackScan
RackScan
RackScan s, code scanner for side reading of SBS racks and cryostorage boxes, accessory for the RackScan product family
Price
1 570,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The RackScan instrument can read up to 96 barcoded vials in parallel.
  • Camera based instrument resulting in rapid imaging and decoding
  • Comfortable and easy plug-in system
  • Small footprint, ideal for benchtop (and integration)
The RackScan can be equipped with the 's' accessory.
Specifications
  • Description:
    RackScan s, code scanner for side reading of SBS racks and cryostorage boxes, accessory for the RackScan product family
  • Seq:
    01