Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
Product Details & Documents
These cassettes are moulded from acetal and keep specimens safely submerged in liquid, and are resistant to the chemical action of most histological solvents.
- Anterior writing area at a 45° angle
- Taped cassettes to be used with Leica ink jet printers
- Easy to remove disposable cover
Unisette™ histology cassette comes with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap-latch and hinge-lock design of the UNISETTE™ prevents early separation of base and lid and allows for one-handed operation.
Packaging: Cassettes are packed in stacks of 40. Each case contains 25 stacks for a total of 1000 cassettes.
Specifications
- Colour:Turquoise
- Seq:13



