Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T

Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T

Green
Catalog # SIMPM525-4T
Supplier:   Simport Scientific
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
Catalog # SIMPM525-4T
Supplier:  Simport Scientific
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T

Green

Price
241,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

These cassettes are moulded from acetal and keep specimens safely submerged in liquid, and are resistant to the chemical action of most histological solvents.

  • Anterior writing area at a 45° angle
  • Taped cassettes to be used with Leica ink jet printers
  • Easy to remove disposable cover

Unisette™ histology cassette comes with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.

The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap-latch and hinge-lock design of the UNISETTE™ prevents early separation of base and lid and allows for one-handed operation.

Packaging: Cassettes are packed in stacks of 40. Each case contains 25 stacks for a total of 1000 cassettes.

Specifications
  • Colour:
    Green
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.