Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T

Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T

White
Catalog # SIMPM525-2T
Supplier:   Simport Scientific
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
Catalog # SIMPM525-2T
Supplier:  Simport Scientific
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T

White

Price
241,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

These cassettes are moulded from acetal and keep specimens safely submerged in liquid, and are resistant to the chemical action of most histological solvents.

  • Anterior writing area at a 45° angle
  • Taped cassettes to be used with Leica ink jet printers
  • Easy to remove disposable cover

Unisette™ histology cassette comes with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.

The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap-latch and hinge-lock design of the UNISETTE™ prevents early separation of base and lid and allows for one-handed operation.

Packaging: Cassettes are packed in stacks of 40. Each case contains 25 stacks for a total of 1000 cassettes.

Specifications
  • Colour:
    White
  • Seq:
    01