Containers with Sloping Base
90° bend, —, Stainless steel 316L
Catalog # 300-0642
Supplier: Sampling Systems
Jump to:
Product Details & Documents
These drums which incorporate a sloping base allow the container to drain completely.
- Made from 316L stainless steel
- Includes lid, clamp and silicone seal
- Other seals available
- Ferrule for easy emptying
- Handles enable easy transportation
The Sampling Systems range of containers with sloping base ensures that material can be completely drained from within and include a ferrule at the base for easy emptying. They have a mirror polish finish for easy cleaning.
Specifications
- Capacity:—
- Description:90° bend
- Material:Stainless steel 316L
- Seq:04
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



