Order Entry
Belgium
Orders LinkContactUsLinkComponent
Blower transition adapters

Blower transition adapters

Blower transition adapter, 152,4 mm nominal diameter, 1,3 kg
Catalog # LBCP4722401
Supplier:   LABCONCO
Blower transition adapters
Catalog # LBCP4722401
Supplier:  LABCONCO
Blower transition adapters
Blower transition adapters

Blower transition adapter, 152,4 mm nominal diameter, 1,3 kg

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Epoxy-coated steel blower transition adapters fit all Labconco coated steel blowers.

Adapter allows you to connect thermoplastic duct to the exhaust side of a blower to create an exhaust stack. A nominal size PVC duct fits inside the opening.

Specifications
  • Description:
    Blower transition adapter, 152,4 mm nominal diameter
  • Seq:
    01
  • Weight:
    1,3 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.