Orders LinkContactUsLinkComponent
Objectives
Objectives
HI PLAN phase objective PH1, 0,92 mm
Catalogus # LEIM11506278
Leverancier:   LEICA MICROSYSTEMS
Objectives
Catalogus # LEIM11506278
Leverancier:  LEICA MICROSYSTEMS
Objectives
HI PLAN phase objective PH1, 0,92 mm
Price
413,87 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Plan achromatic and phase contrast objectives.
Specificaties
  • Magnification:
    20×/0,40
  • Description:
    HI PLAN phase objective PH1
  • Working distance:
    0,92 mm