Objectives
HI PLAN achromatic objective, 0,35 mm
LEIM11506238
Plan achromatic and phase contrast objectives.
- Magnification:40×/0,65
- Description:HI PLAN achromatic objective
- Working distance:0,35 mm
Plan achromatic and phase contrast objectives.